Få mere at vide
Cytokeratin 18 Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
Brand: Epredia MS-142-B
Beskrivelse
Keratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with keratin 8. It was reported that tissues from gastrointestinal tract are positive for both keratin 8 and 18 but do not contain keratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for keratin 18.Host Species: Mouse
Clone: DC10
Isotype: IgG1
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Human breast cancer PMC 42 cells
Molecular Weight: 45 kDa
Positive Control: HT29 cells, Skin, Breast or Lung CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Tekniske data
| Cytokeratin 18 Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 1 mL | |
| Cytoskeletal Protein | |
| Human | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| DC10 | |
| Biotin | |
| Human breast cancer PMC 42 cells | |
| IVD | |
| Primary | |
| Concentrated |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.