Learn More
CD45RO (T-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Epredia™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody.
Brand:  Epredia MS-112-P
Additional Details : Weight : 0.00250kg
 View more versions of this product
Product Code. 12679056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
CD45RO (an isoform of leukocyte common antigen) is expressed on most thymocytes, monocytes, macrophages and granulocytes. It is not expressed on normal B lymphocytes. Intrathymic T cell maturation progresses from CD45RA-/CD45RO+ to CD45RA+/CD45RO-; while peripheral T cell maturation progresses from CD45RA+/CD45RO- to CD45RA-/CD45RO+.
Host Species: Mouse
Clone: UCHL-1
Isotype: IgG2a/κ
Species Reactivity: Human, Rhesus Monkey and Chimpanzee. Does not react with rat. Others not known
Epitope: Not determined
Immunogen: Interleukin-2-dependent human T lymphocytes
Molecular Weight: 180 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
Specifications
Specifications
CD45RO (T-Cell Marker) Ab-1
Monoclonal
100ug/mL
Mouse
1 mL
Oncogenesis
Human, Non-human Primate
IgG2a κ
Flow Cytometry, Immunohistochemistry (Paraffin)
UCHL-1
Unconjugated
Interleukin-2-dependent human T lymphocytes
IVD
Primary
Concentrated