Få mere at vide
STEMCELL Technologies L755507
Beskrivelse
- L755507 is a potent and selective β-3 adrenergic receptor agonist that demonstrates > 440-fold selectivity over β-1 and β-2 binding (EC50 = 0.43, 580, and > 10,000 nM for activation of cloned human β-3-, β-2-, and β-1-adrenoreceptors, respectively; Parmee et al.)
- Stimulates lipolysis in human white adipose tissue (Fisher et al.)
- Improves the efficiency of homology-directed repair in human induced-pluripotent stem cells following CRISPR editing (Yu et al.)
- Increases metabolism in rhesus monkeys (Fisher et al.)
Tekniske data
Tekniske data
| CAS | 159182-43-1 |
| Mængde | 10 mg |
| Molekylær formel | C30H40N4O6S |
| Renhed | ≥98% |