Få mere at vide
L-Valine Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-Valine Methyl Ester Hydrochloride |
| CAS | 6306-52-1 |
| Molekylær formel | C6 H13 N O2 . Cl H |
| Mængde | 25 g |
| Synonym | L-Valine','methyl ester','hydrochloride (1:1), L-Valine','methyl ester','hydrochloride (9CI), Valine','methyl ester','hydrochloride','L- (6CI,7CI,8CI), (S)-Valine methyl ester hydrochloride, Methyl (S)-2-amino-3-methylbutanoate hydrochloride, Methyl (S)-valinate hydrochloride, Methyl L-valinate hydrochloride, Methyl valinate hydrochloride, NSC 197198, NSC 22920, Valine methyl ester hydrochloride |
| InChI formel | InChI=1S/C6H13NO2.ClH/c1-4(2)5(7)6(8)9-3;/h4-5H,7H2,1-3H3;1H/t5-;/m0./s1 |
| SMIL | Cl.COC(=O)[C@@H](N)C(C)C |
| IUPAC navn | methyl (2S)-2-amino-3-methylbutanoate;hydrochloride |
| Molekylvægt (g/mol) | 167.63 |
| Formel vægt | 167.0713 |