Få mere at vide
L-Valine-2-d1, TRC
Artikelnummer. 30455223
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30455223 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455223 Leverandør Toronto Research Chemicals Leverandørnr. TRCV09420650MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Valine-2-d1
CAS 77257-03-5
Molekylær formel C5 D H10 N O2
Mængde 50 mg
Synonym L-Valine-2-d (9CI','ACI), L-(α-2H)Valine, (2S)-2-Amino-3-methylbutanoic-2-d acid, L-Valine-2-D
InChI formel InChI=1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i4D
SMIL [2H][C@](N)(C(C)C)C(=O)O
IUPAC navn (2S)-2-amino-2-deuterio-3-methylbutanoic acid
Molekylvægt (g/mol) 118.153
Formel vægt 118.085