Få mere at vide
L-Valine-13C5,15N, TRC
Artikelnummer. 30136689
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136689 50 mg
30011648 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136689 Leverandør Toronto Research Chemicals Leverandørnr. TRCV09420750MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202407-30-5
Molekylær formel 13C5H1115NO2
Molekylvægt (g/mol) 123.1
Synonym (+)-2-Amino-3-methylbutyric Acid-13C5,15N, (2S)-2-Amino-3-methylbutanoic Acid-13C5,15N, (S)-2-Amino-3-methylbutanoic Acid-13C5,15N, (S)-Valine-13C5,15N, (S)-α-Amino-β-methylbutyric Acid-13C5,15N, L-(+)-α-Aminoisovaleric Acid-13C5,15N, L-α-Amino-β-methylbutyric Acid-13C5,15N, NSC 76038-13C5,15N;
SMIL [15NH2][13C@@]([13C](O)=O)([H])[13CH]([13CH3])[13CH3]
Tekniske data
Kemisk navn eller materiale L-Valine-13C5,15N
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 123.0928
Procent renhed >95%