Få mere at vide
L-Tyrosine Ethyl Ester, Mikromol™
Artikelnummer. 30051077
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051077 25 mg
30050416 100 mg
30463208 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051077 Leverandør Mikromol Leverandørnr. MM0634.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
L-Tyrosine Ethyl Ester
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Tyrosine Ethyl Ester
CAS 949-67-7
Analyt- eller komponentnavne L-Tyrosine Ethyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C11 H15 N O3
Certificeringer/compliance ISO 17025
Synonym L-Tyrosine ethyl ester, Tyrosine, ethyl ester, L- (6CI,8CI), (S)-Tyrosine ethyl ester, Ethyl (S)-2-amino-3-(4-hydroxyphenyl)propionate, Ethyl L-tyrosinate, NSC 519993
InChI formel InChI=1S/C11H15NO3/c1-2-15-11(14)10(12)7-8-3-5-9(13)6-4-8/h3-6,10,13H,2,7,12H2,1H3/t10-/m0/s1
SMIL CCOC(=O)[C@@H](N)Cc1ccc(O)cc1
IUPAC navn ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Molekylvægt (g/mol) 209.242
Mængde 25 mg
Formel vægt 209.105 g/mol