Få mere at vide
L-Tyrosine-Alpha,Beta,Beta,2,3,5,6-d7, TRC
Artikelnummer. 30136070
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136070 5 mg
30134000 1 mg
17927769 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30136070 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8999955MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 130551-49-4
Molekylær formel C9 D7 H4 N O3
Molekylvægt (g/mol) 188.232
Synonym L-Tyrosine-α,β,β,2,3,5,6-d7 (9CI, ACI), L-Tyrosine-D7, (2S)-2-Amino-3-(4-hydroxyphenyl-2,3,5,6-d4)propanoic-2,3,3-d3 acid, 4-Hydroxyphenyl-d4-L-alanine-2,3,3-d3, β-(p-Hydroxyphenyl-d4)alanine-α,β,β-d3, L-Tyrosine-d7, L-4-Hydroxyphenylalanine-d7
IUPAC navn (2S)-2-amino-2,3,3-trideuterio-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)propanoic acid
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1O)C([2H])([2H])[C@]([2H])(N)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Tyrosine-alpha,beta,beta,2,3,5,6-d7
InChI formel InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1/i1D,2D,3D,4D,5D2,8D
Mængde 5 mg
Formel vægt 188.118