Få mere at vide
L-Tyrosine-13C9,15N, TRC
Artikelnummer. 30006084
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006084 5 mg
30011436 100 mg
30011507 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006084 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8999765MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202407-26-9
Molekylær formel 13C9 H11 15N O3
Molekylvægt (g/mol) 191.1158
Synonym L-Tyrosine-carboxy,α,β,1,2,3,4,5,6-13C9-15N, L-Tyrosine-13C9-15N (9CI), L-[13C9,15N]-Tyrosine, (2S)-2-(Amino-15N)-3-(4-hydroxyphenyl-1,2,3,4,5,6-13C6)propanoic-1,2,3-13C3 acid, 4-Hydroxyphenyl-13C6-L-alanine-13C3-15N, β-(p-Hydroxyphenyl)alanine-13C9,15N, L-Tyrosine-13C9,15N
IUPAC navn (2S)-2-(^{15}N)azanyl-3-(4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)(1,2,3-^{13}C_{3})propanoic acid
SMIL [15NH2][13C@@H]([13CH2][13c]1[13cH][13cH][13c](O)[13cH][13cH]1)[13C](=O)O
Tekniske data
Kemisk navn eller materiale L-Tyrosine-13C9-15N
Mængde 5 mg
InChI formel InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1
Formel vægt 191.1011