Få mere at vide
L-Tryptophan Isopropyl Ester Hydrochloride, Mikromol™
Artikelnummer. 30050945
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050945 100 mg
30049212 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050945 Leverandør Mikromol Leverandørnr. MM0172.19
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
L-Tryptophan Isopropyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Tryptophan Isopropyl Ester Hydrochloride
CAS 71959-71-2
Analyt- eller komponentnavne L-Tryptophan Isopropyl Ester Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C14 H18 N2 O2 . Cl H
Certificeringer/compliance ISO 9001
Synonym L-Tryptophan Isopropyl Ester Hydrochloride, L-Tryptophan, 1-methylethyl ester, hydrochloride (1:1), L-Tryptophan, 1-methylethyl ester, monohydrochloride (9CI)
InChI formel InChI=1S/C14H18N2O2.ClH/c1-9(2)18-14(17)12(15)7-10-8-16-13-6-4-3-5-11(10)13;/h3-6,8-9,12,16H,7,15H2,1-2H3;1H/t12-;/m0./s1
SMIL Cl.CC(C)OC(=O)[C@@H](N)Cc1c[nH]c2ccccc12
IUPAC navn propan-2-yl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Molekylvægt (g/mol) 282.77
Mængde 100 mg
Formel vægt 282.1135 g/mol