Få mere at vide
L-Tryptophan-d5, TRC
Artikelnummer. 30454314
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30454314 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454314 Leverandør Toronto Research Chemicals Leverandørnr. TRCT9472122.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Tryptophan-d5
CAS 62595-11-3
Renhedsgrad noter HPLC
Molekylær formel C11 2H5 H7 N2 O2
Mængde 2.5 mg
Synonym L-Tryptophan-2,4,5,6,7-D5, L-(2,4,5,6,7-2H5)Tryptophan, L-Tryptophan-d5, (S)-Tryptophan-d5, (S)-α-Amino-3-(indole-2,4,5,6,7-d5)propionic acid, (S)-α-Amino(indole-2,4,5,6,7-d5)-3-propanoic acid
InChI formel InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1/i1D,2D,3D,4D,6D
SMIL [2H]c1[nH]c2c([2H])c([2H])c([2H])c([2H])c2c1C[C@H](N)C(=O)O
IUPAC navn (2S)-2-amino-3-(2,4,5,6,7-pentadeuterio-1H-indol-3-yl)propanoic acid
Molekylvægt (g/mol) 209.26
Formel vægt 209.1213
Procent renhed >95