missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, 98.5 to 101.5% (Dry Basis), Thermo Scientific™

Catalog Number p-7082828
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
73-22-3
C11H12N2O2
204.23
MFCD00064340
QIVBCDIJIAJPQS-VIFPVBQESA-N
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
6305
CHEBI:16828
(2 S)-2-amino-3-(1 H-indol-3-yl)propanoic acid
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
73-22-3
C11H12N2O2
204.23
MFCD00064340
QIVBCDIJIAJPQS-VIFPVBQESA-N
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
6305
CHEBI:16828
(2 S)-2-amino-3-(1 H-indol-3-yl)propanoic acid
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Melting Point 280°C to 285°C
Beilstein 86197
Packaging Plastic Bottle
Formula Weight 204.23
Percent Purity 98.5 to 101.5%
Quantity 100 g
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Tryptophan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'