Få mere at vide
L-Tryptophan-2,3,3-d3, CDN
Artikelnummer. 30313947
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313947 0.05 g
30316090 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313947 Leverandør CDN Leverandørnr. CDND74190.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Tryptophan-d3
CAS 133519-78-5
Molekylær formel C11 D3 H9 N2 O2
Mængde 0.05 g
Synonym L-Tryptophan-α,β,β-d3 (9 CI), α,β,β-Tri-2 H-L-tryptophan, L-Tryptophan-α,β,β-d3, L-Tryptophan-d3, (S)-α-Amino-3-indole-α,β,β-d3-propionic acid, (αS)-α-Aminoindole-3-propanoic-d3 acid, L-Tryptophan-α,β,β-D3
InChI formel InChI=1 S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13 H,5,12H2,(H,14,15)/t9-/m0/s1/i5D2,9 D
SMIL [2 H]C([2 H])(c1c[nH]c2ccccc12)[C@]([2 H])(N)C(=O)O
IUPAC navn (2 S)-2-amino-2,3,3-trideuterio-3-(1 H-indol-3-yl)propanoic acid
Molekylvægt (g/mol) 207.244
Formel vægt 207.109 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity