Få mere at vide
L-Thyronine, 95%, TRC
Artikelnummer. 17960755
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17960755 100 mg
17994685 250 mg
17972855 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17960755 Leverandør Toronto Research Chemicals Leverandørnr. TRCT922700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1596-67-4
Molekylær formel C15 H15 N O4
Molekylvægt (g/mol) 273.28
Synonym L-Thyronine, 95%, O-(4-Hydroxyphenyl)-L-tyrosine, Desiodothyroxine, β-(p-Hydroxyphenoxy)phenylalanine
IUPAC navn (2S)-2-amino-3-[4-(4-hydroxyphenoxy)phenyl]propanoic acid
SMIL N[C@@H](Cc1ccc(Oc2ccc(O)cc2)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Thyronine
InChI formel InChI=1S/C15H15NO4/c16-14(15(18)19)9-10-1-5-12(6-2-10)20-13-7-3-11(17)4-8-13/h1-8,14,17H,9,16H2,(H,18,19)/t14-/m0/s1
Mængde 100 mg
Formel vægt 273.1001