Få mere at vide
L-threo-Phenylserine, TRC
Artikelnummer. 30423663
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423663 500 mg
30409580 250 mg
30410160 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423663 Leverandør Toronto Research Chemicals Leverandørnr. TRCP336833500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6254-48-4
Molekylær formel C9H11NO3
Molekylvægt (g/mol) 181.19
Synonym (betaR)-beta-Hydroxy-L-phenylalanine, (2S,3R)-2-Amino-3-hydroxy-3-phenylpropanoic Acid, (2S,3R)-2-Amino-3-hydroxy-3-phenylpropionic Acid, L-threo-3-Phenylserine, L-threo-beta-Phenylserine, threo-beta-Hydroxy-L-phenylalanine
IUPAC navn (2S,3R)-2-amino-3-hydroxy-3-phenylpropanoic acid
SMIL N[C@@H]([C@H](O)c1ccccc1)C(=O)O
Tekniske data
Kemisk navn eller materiale L-threo-Phenylserine
InChI formel InChI=1S/C9H11NO3/c10-7(9(12)13)8(11)6-4-2-1-3-5-6/h1-5,7-8,11H,10H2,(H,12,13)/t7-,8+/m0/s1
Mængde 500 mg
Formel vægt 181.0739