Få mere at vide
L-Tetrahydrofolic Acid-d4 (Major) (>70% when packaged), TRC
Artikelnummer. 30413924
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30413924 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413924 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2935171MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Tetrahydrofolic Acid-d4 (Major)
CAS 1356841-72-9
Molekylær formel C19H19D4N7O6
Mængde 1 mg
Synonym N-[4-[[(2-Amino-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic Acid-d4, (-)-L-5,6,7,8-Tetrahydrofolic Acid-d4, THFA-d4, Tetrahydropteroylglutamic Acid-d4
InChI formel InChI=1S/C19H23N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,11-12,21,23H,5-8H2,(H,24,29)(H,27,28)(H,31,32)(H4,20,22,25,26,30)/t11?,12-/m0/s1/i7D2,8D,11D/t8?,11?,12-
SMIL [2H]C1NC2=C(NC1([2H])C([2H])([2H])Nc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)C(=O)N=C(N)N2
IUPAC navn (2S)-2-[[4-[[(2-amino-6,7-dideuterio-4-oxo-1,5,7,8-tetrahydropteridin-6-yl)-dideuteriomethyl]amino]benzoyl]amino]pentanedioic acid
Molekylvægt (g/mol) 449.45
Formel vægt 449.1961