Få mere at vide
L-Serine-d7, TRC
Artikelnummer. 30419956
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30419956 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419956 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2710061MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Serine-d7
CAS 935275-35-7
Molekylær formel C3 D7 N O3
Mængde 1 mg
Synonym L-Serine-N,N,O,1,2,3,3-d7 (ACI), L-Serine-d7, L-Serine-D7, (2S)-2-(Amino-d2)-3-(hydroxy-d)propanoic-2,3,3-d3 acid-d
InChI formel InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1/i1D2,2D,5D/hD3
SMIL [2H]OC(=O)[C@@]([2H])(N([2H])[2H])C([2H])([2H])O[2H]
IUPAC navn deuterio (2S)-2,3,3-trideuterio-3-deuteriooxy-2-(dideuterioamino)propanoate
Molekylvægt (g/mol) 112.1357
Formel vægt 112.0865