Få mere at vide
L-Serine-d7, CDN
Artikelnummer. 30325601
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325601 0.05 g -
30311834 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325601 Leverandør CDN Leverandørnr. CDND64660.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Serine-d7
CAS 935275-35-7
Molekylær formel C3 D7 N O3
Mængde 0.05 g
Synonym L-Serine-N,N,O,1,2,3,3-d7 (ACI), L-Serine-d7, L-Serine-D7, (2 S)-2-(Amino-d2)-3-(hydroxy-d)propanoic-2,3,3-d3 acid-d
InChI formel InChI=1 S/C3H7NO3/c4-2(1-5)3(6)7/h2,5 H,1,4H2,(H,6,7)/t2-/m0/s1/i1D2,2 D,5 D/hD3
SMIL [2 H]OC(=O)[C@@]([2 H])(N([2 H])[2 H])C([2 H])([2 H])O[2 H]
IUPAC navn deuterio (2 S)-2,3,3-trideuterio-3-deuteriooxy-2-(dideuterioamino)propanoate
Molekylvægt (g/mol) 112.1357
Formel vægt 112.0865 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity