Få mere at vide
L-Serine-13C, TRC
Artikelnummer. 30421089
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421089 25 mg
30409215 5 mg
30414755 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421089 Leverandør Toronto Research Chemicals Leverandørnr. TRCS27106825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81201-84-5
Molekylær formel C213CH7NO3
Molekylvægt (g/mol) 106.09
Synonym [1-13C]-Serine
IUPAC navn (2S)-2-amino-3-hydroxy(113C)propanoic acid
SMIL N[C@@H](CO)[13C](=O)O
Tekniske data
Kemisk navn eller materiale L-Serine-13C
InChI formel InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1/i3+1
Mængde 25 mg
Formel vægt 106.0459