missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Selectin/CD62L Antibody (MEL-14), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81083APC
Additional Details : Weight : 0.00970kg
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Flow Cytometry.Specifications
L-Selectin/CD62L | |
Monoclonal | |
APC | |
PBS | |
SELL | |
C3H/eb mouse B lymphoma 38C-13. | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cardiovascular Biology, Glycobiology, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cells | |
6402 | |
Mouse | |
Purified |
Flow Cytometry | |
MEL-14 | |
Flow Cytometry | |
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds to mouse L-Selectin/CD62L the mouse peripheral lymph node homing receptor, which is a lectin-like cell surface protein rapidly down-regulated upon cell activation with phorbol 12-myristate 13-acetate. | |
Store at 4°C in the dark. | |
IgG κ |