Få mere at vide
L-Saccharopine, TRC
Artikelnummer. 30011924
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011924 5 mg
30011670 10 mg
30003278 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011924 Leverandør Toronto Research Chemicals Leverandørnr. TRCS8083405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 997-68-2
Molekylær formel C11 H20 N2 O6
Molekylvægt (g/mol) 276.29
Synonym Glutamic acid, N-(5-amino-5-carboxypentyl)-, L- (8CI), L-Glutamic acid, N-(5-amino-5-carboxypentyl)-, (S)-, Saccharopine (7CI), N-[(5S)-5-Amino-5-carboxypentyl]-L-glutamic acid, L-Saccharopine, Saccharopin
IUPAC navn (2S)-2-[[(5S)-5-amino-5-carboxypentyl]amino]pentanedioic acid
SMIL N[C@@H](CCCCN[C@@H](CCC(=O)O)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Saccharopine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C11H20N2O6/c12-7(10(16)17)3-1-2-6-13-8(11(18)19)4-5-9(14)15/h7-8,13H,1-6,12H2,(H,14,15)(H,16,17)(H,18,19)/t7-,8-/m0/s1
Mængde 5 mg
Formel vægt 276.1321
Procent renhed >95%