Få mere at vide
L-Ribulose, TRC
Artikelnummer. 30413680
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413680 50 mg
30415539 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30413680 Leverandør Toronto Research Chemicals Leverandørnr. TRCR41800350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2042-27-5
Molekylær formel C5 H10 O5
Molekylvægt (g/mol) 150.13
Synonym L-erythro-Pentulose
IUPAC navn (3S,4S)-1,3,4,5-tetrahydroxypentan-2-one
SMIL OC[C@H](O)[C@H](O)C(=O)CO
Tekniske data
Kemisk navn eller materiale L-Ribulose
Mængde 50 mg
InChI formel InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h3,5-8,10H,1-2H2/t3-,5-/m0/s1
Formel vægt 150.0528