Få mere at vide
L-Rhamnitol, TRC
Artikelnummer. 30415066
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415066 500 mg
30428130 1 g
30420148 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30415066 Leverandør Toronto Research Chemicals Leverandørnr. TRCR314990500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 488-28-8
Molekylær formel C6H14O5
Molekylvægt (g/mol) 166.17
Synonym 1-Deoxy-L-mannitol, 6-Deoxy-L-mannitol, L-Rhamnitol, Rhamnitol
IUPAC navn (2S,3S,4S,5S)-hexane-1,2,3,4,5-pentol
SMIL C[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)CO
Tekniske data
Kemisk navn eller materiale L-Rhamnitol
Mængde 500 mg
InChI formel InChI=1S/C6H14O5/c1-3(8)5(10)6(11)4(9)2-7/h3-11H,2H2,1H3/t3-,4-,5-,6-/m0/s1
Formel vægt 166.0841