Få mere at vide
L-Pyroglutamic Acid-13C5, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-Pyroglutamic Acid-13C5 |
| CAS | 55443-56-6 |
| Molekylær formel | 13C5H7NO3 |
| Mængde | 10 mg |
| Synonym | (-)-2-Pyrrolidone-5-carboxylic Acid-13C5, (-)-Pyroglutamic Acid-13C5, (2S)-5-Oxopyrrolidine-2-carboxylic Acid-13C5, (5S)-2-Oxopyrrolidine-5-carboxylic Acid-13C5, (S)-(-)-2-Pyrrolidone-5-carboxylic Acid-13C5, (S)-(-)-γ-Butyrolactam-γ-carboxylic Acid-13C5, (S)-2-Pyrrolidone-5-carboxylic Acid-13C5, (S)-5-Oxo-2-pyrrolidinecarboxylic Acid-13C5, (S)-5-Oxoproline-13C5, (S)-Pyroglutamic Acid-13C5, 2-L-Pyrrolidone-5-carboxylic Acid-13C5, 2-Oxopyrrolidine-5(S)-carboxylic Acid, -13C5 2-Pyrrolidinone-5-carboxylic Acid-13C5, 5-Carboxy-2-pyrrolidinone-13C5, 5-Oxo-L-proline-13C5, 5-Oxoproline-13C5, 5-Pyrrolidinone-2-carboxylic Acid-13C5, Ajidew A 100-13C5, Glutimic Acid-13C5, Glutiminic Acid-13C5, L-(-)-Pyroglutamic Acid-13C5, L-2-Pyrrolidone-5-carboxylic Acid-13C5, L-5-Carboxy-2-pyrrolidinone-13C5, L-5-Oxo-2-pyrrolidinecarboxylic Acid-13C5, L-5-Oxoproline-13C5, γ-Lactam L-Glutamic Acid-13C5, L-Glutimic Acid-13C5, L-Glutiminic Acid, -13C5 L-Pyrrolidinonecarboxylic Acid-13C5, L-Pyrrolidonecarboxylic Acid-13C5, NSC 143034-13C5, Oxoproline-13C5, Oxopyrrolidinecarboxylic Acid-13C5, PCA-13C5, PCA 50-13C5, Pidolic Acid-13C5, Pidolidone-13C5, Pyroglutamic Acid-13C5, Pyrrolidinonecarboxylic Acid-13C5, Pyrrolidone-5-carboxylic Acid-13C5, Pyrrolidonecarboxylic Acid-13C5 |
| InChI formel | InChI=1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1/i1+1,2+1,3+1,4+1,5+1 |
| SMIL | O=[13C]1[13CH2][13CH2][13C@]([H])(N1)[13C](O)=O |
| IUPAC navn | (S)-5-oxopyrrolidine-2-carboxylic-2,3,4,5-13C4 acid-13C |
| Molekylvægt (g/mol) | 134.08 |
| Formel vægt | 134.06 |