Learn More
L-Proline methyleester hydrochloride, 98%, Thermo Scientific Chemicals
Catalog Number p-3757516
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
2133-40-6
C6H12NO2
130.17
MFCD00012708
BLWYXBNNBYXPPL-YFKPBYRVSA-O
l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride
2733200
methyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
COC(=O)[C@@H]1CCC[NH2+]1
2133-40-6
C6H12NO2
130.17
MFCD00012708
BLWYXBNNBYXPPL-YFKPBYRVSA-O
l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride
2733200
methyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
COC(=O)[C@@H]1CCC[NH2+]1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation -31° to -35° (20°C, 589nm) (c=1, H2O)
Formula Weight 165.62
Percent Purity ≥97.5%
Water 0.5% max. (K.F.)
Specific Rotation Condition −33° (20°C c=1,H2O)
Quantity 1 g
Physical Form Crystalline Powder, Crystals, and/or Chunks
Chemical Name or Material L-Proline methyleester hydrochloride