Få mere at vide
L-Pipecolic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-Pipecolinic Acid |
| CAS | 3105-95-1 |
| Molekylær formel | C6 H11 N O2 |
| Synonym | 2-Piperidinecarboxylic acid, (2S)-, 2-Piperidinecarboxylic acid, (S)-, Pipecolic acid, (S)-(-)- (8CI), (2S)-2-Piperidinecarboxylic acid, (-)-Pipecolic acid, (-)-Pipecolinic acid, (2S)-Piperidin-1-ium-2-carboxylate, (2S)-Piperidine-2-carboxylic acid, (S)-(-)-2-Piperidinecarboxylic acid, (S)-(-)-Pipecolic acid, (S)-Pipecolic acid, (S)-Pipecolinic acid, (S)-Piperidine-2-carboxylic acid, 2-Piperidinecarboxylic acid (2S)-, L-(-)-Pipecolic acid, L-Homoproline, L-Pipecolic acid, L-Pipecolinic acid, L-Piperidine-2-carboxylic acid, NSC 93089 |
| InChI formel | InChI=1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)/t5-/m0/s1 |
| SMIL | OC(=O)[C@@H]1CCCCN1 |
| IUPAC navn | (2S)-piperidine-2-carboxylic acid |
| Molekylvægt (g/mol) | 129.16 |
| Mængde | 100 mg |
| Formel vægt | 129.079 |
| Vis mere |