Få mere at vide
L-Phenylglycine Amide, TRC
Artikelnummer. 30012692
Skift visning
Mængde:
25 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012692 50 g
30009358 250 g
17924245 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012692 Leverandør Toronto Research Chemicals Leverandørnr. TRCP32131050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6485-52-5
Molekylær formel C8 H10 N2 O
Molekylvægt (g/mol) 150.18
Synonym Benzeneacetamide, α-amino-, (αS)- (9CI, ACI), (αS)-α-Aminobenzeneacetamide (ACI), Acetamide, 2-amino-2-phenyl-, L-(+)- (8CI), Benzeneacetamide, α-amino-, (S)- (ZCI)
IUPAC navn (2S)-2-amino-2-phenylacetamide
SMIL N[C@H](C(=O)N)c1ccccc1
Tekniske data
Kemisk navn eller materiale L-Phenylglycine Amide
InChI formel InChI=1S/C8H10N2O/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H2,10,11)/t7-/m0/s1
Mængde 50 g
Formel vægt 150.0793