Få mere at vide
L-Phenylglycine Amide, Mikromol™
Beskrivelse
L-Phenylglycine Amide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-Phenylglycine Amide |
| CAS | 6485-52-5 |
| Analyt- eller komponentnavne | L-Phenylglycine Amide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H10 N2 O |
| Certificeringer/compliance | ISO 9001 |
| Synonym | L-(+)-2-Amino-2-phenyl-acetamide, (S)-α-Amino-benzeneacetamide, (2S)-2-Amino-2-phenylethanamide, L-Phenylglycinamide, L-α-Phenylglycine amide, Benzeneacetamide, α-amino-, (αS)- (9CI, ACI), (αS)-α-Aminobenzeneacetamide (ACI), Acetamide, 2-amino-2-phenyl-, L-(+)- (8CI), Benzeneacetamide, α-amino-, (S)- (ZCI) |
| InChI formel | InChI=1S/C8H10N2O/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H2,10,11)/t7-/m0/s1 |
| SMIL | N[C@H](C(=O)N)c1ccccc1 |
| IUPAC navn | (2S)-2-amino-2-phenylacetamide |
| Vis mere |