Få mere at vide
L-Phenylalanine tert-Butyl Ester-d5, TRC
Artikelnummer. 30411985
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30411985 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411985 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3194421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Phenylalanine tert-Butyl Ester-d5
CAS 1177315-89-7
Molekylær formel C13H14D5NO2
Mængde 1 mg
Synonym Phenylalanine tert-Butyl Ester-d5, tert-Butyl (S)-2-Amino-3-phenylpropanoate-d5, tert-Butyl L-Phenylalaninate-d5, (S)-2-Amino-3-phenylpropionic Acid tert-Butyl Ester-d5, L-Phenylalanine','1,1-Dimethylethyl Ester-d5
InChI formel InChI=1S/C13H19NO2/c1-13(2,3)16-12(15)11(14)9-10-7-5-4-6-8-10/h4-8,11H,9,14H2,1-3H3/t11-/m0/s1/i4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(C[C@H](N)C(=O)OC(C)(C)C)c([2H])c1[2H]
IUPAC navn tert-butyl (2S)-2-amino-3-(2,3,4,5,6-pentadeuteriophenyl)propanoate
Molekylvægt (g/mol) 226.33
Formel vægt 226.173