Få mere at vide
L-Phenylalanine-d5, TRC
Artikelnummer. 30423626
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30423626 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423626 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31941810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Phenylalanine-D5
CAS 56253-90-8
Renhedsgrad noter HPLC
Molekylær formel C9 2H5 H6 N O2
Mængde 10 mg
Synonym L-Phenylalanine-(phenyl-D5), L-Phenylalanine-2,3,4,5,6-d5, L-Phenyl-d5-alanine (9CI), L-(2,3,4,5,6-2H5)Phenylalanine, L-Phenyl-2,3,4,5,6-d5-alanine, L-Phenylalanine-d5, [2H5]Phenylalanine, d5-ring-L-Phenylalanine
InChI formel InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C[C@H](N)C(=O)O)c([2H])c1[2H]
IUPAC navn (2S)-2-amino-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid
Molekylvægt (g/mol) 170.22
Formel vægt 170.1104
Procent renhed >95