missing translation for 'onlineSavingsMsg'
Få mere at vide

L-Phenylalanine-d5 Methyl Ester Hydrochloride, TRC

Artikelnummer. 30424588
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424588 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424588 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31942810MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale L-Phenylalanine-d5 Methyl Ester Hydrochloride
CAS 1331897-23-4
Molekylær formel C10H9D5ClNO2
Mængde 10 mg
Synonym (S)-Phenylalanine-d5 Methyl Ester Hydrochloride, Methyl (S)-Phenylalaninate-d5 Hydrochloride, Methyl L-Phenylalaninate-d5 Hydrochloride, NSC 194642-d5
InChI formel InChI=1S/C10H13NO2.ClH/c1-13-10(12)9(11)7-8-5-3-2-4-6-8;/h2-6,9H,7,11H2,1H3;1H/t9-;/m0./s1/i2D,3D,4D,5D,6D;
SMIL O=C([C@@H](N)CC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])OC.Cl
Molekylvægt (g/mol) 220.71
Formel vægt 220.1027
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.