Få mere at vide
L-Phenyl-d5-alanine-2,3,3-d3, TRC
Artikelnummer. 30424924
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30424924 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424924 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31942250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Phenyl-d5-alanine-2,3,3-d3
CAS 17942-32-4
Molekylær formel C9 D8 H3 N O2
Mængde 50 mg
Synonym L-Phenyl-2,3,4,5,6-d5-alanine-α,β,β-d3, L-Phenylalanine-d8
InChI formel InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1/i1D,2D,3D,4D,5D,6D2,8D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])[C@]([2H])(N)C(=O)O
IUPAC navn (2S)-2-amino-2,3,3-trideuterio-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid
Molekylvægt (g/mol) 173.24
Formel vægt 173.1292