missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Perillaldehyde, 90%, Thermo Scientific Chemicals

Catalog Number p-180143
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18031-40-8
C10H14O
150.22
MFCD00001543
RUMOYJJNUMEFDD-SNVBAGLBSA-N
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
2724159
CHEBI:86938
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
CC(=C)C1CCC(=CC1)C=O
This item is not returnable. View return policy
18031-40-8
C10H14O
150.22
MFCD00001543
RUMOYJJNUMEFDD-SNVBAGLBSA-N
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
2724159
CHEBI:86938
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
CC(=C)C1CCC(=CC1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 0.9700g/mL
Boiling Point 104°C to 105°C (10.0 mmHg)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 88% min
Packaging Glass bottle
Refractive Index 1.5040 to 1.51
Quantity 5 g
Beilstein 07, 158
Merck Index 14, 7167
Specific Gravity 0.97
Specific Rotation Condition − 115.00 (20.00°C c=10,C2H5OH)
Specific Rotation -115
Formula Weight 150.22
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material L(-)-Perillaldehyde, 90%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Perillaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'