Få mere at vide
L-Ornithine-d6 Hydrochloride, TRC
Artikelnummer. 30411747
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30411747 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411747 Leverandør Toronto Research Chemicals Leverandørnr. TRCO6955532.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Ornithine-d6 Hydrochloride
CAS 347841-40-1
Molekylær formel C5 2H6 H6 N2 O2 . Cl H
Mængde 2.5 mg
Synonym L-Ornithine-3,3,4,4,5,5-d6','hydrochloride (1:1), L-Ornithine-3,3,4,4,5,5-d6','monohydrochloride (9CI), L-Ornithine-D6 Hydrochloride
InChI formel InChI=1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1/i1D2,2D2,3D2;
SMIL Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])[C@H](N)C(=O)O
IUPAC navn (2S)-2,5-diamino-3,3,4,4,5,5-hexadeuteriopentanoic acid;hydrochloride
Molekylvægt (g/mol) 174.66
Formel vægt 174.1042