Få mere at vide
L-O-Phosphoserine, TRC
Artikelnummer. 30135512
Skift visning
Mængde:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135512 10 g
30134232 1 g
30010414 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135512 Leverandør Toronto Research Chemicals Leverandørnr. TRCP36120010G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 407-41-0
Molekylær formel C3 H8 N O6 P
Molekylvægt (g/mol) 185.07
Synonym O-Phosphono-L-serine, L-Serine, dihydrogen phosphate (ester) (9CI), Serine phosphate, L- (6CI), Serine, dihydrogen phosphate (ester), L- (8CI), O-Phosphono-L-serine, 3-O-Phosphoserine, Dexfosfoserine, Fosforina, L-3-Phosphoserine, L-O-Phosphoserine, L-O-Serine phosphate, L-Phosphoserine, L-Serinephosphoric acid, L-Seryl phosphate, O-Phospho-L-serine, O-Phosphoryl-L-serine, O-Phosphorylserine, Phospho-L-serine, Serine O-phosphate, Serine dihydrogen phosphate (ester), Seriphos
IUPAC navn (2S)-2-amino-3-phosphonooxypropanoic acid
SMIL N[C@@H](COP(=O)(O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale L-O-Phosphoserine
InChI formel InChI=1S/C3H8NO6P/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)/t2-/m0/s1
Mængde 10 g
Formel vægt 185.0089