missing translation for 'onlineSavingsMsg'
Få mere at vide

L-O-Phosphoserine, TRC

Artikelnummer. 30134232
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134232 1 g
30135512 10 g
30010414 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30134232 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3612001G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 407-41-0
Molekylær formel C3 H8 N O6 P
Molekylvægt (g/mol) 185.07
Synonym O-Phosphono-L-serine, L-Serine, dihydrogen phosphate (ester) (9CI), Serine phosphate, L- (6CI), Serine, dihydrogen phosphate (ester), L- (8CI), O-Phosphono-L-serine, 3-O-Phosphoserine, Dexfosfoserine, Fosforina, L-3-Phosphoserine, L-O-Phosphoserine, L-O-Serine phosphate, L-Phosphoserine, L-Serinephosphoric acid, L-Seryl phosphate, O-Phospho-L-serine, O-Phosphoryl-L-serine, O-Phosphorylserine, Phospho-L-serine, Serine O-phosphate, Serine dihydrogen phosphate (ester), Seriphos
IUPAC navn (2S)-2-amino-3-phosphonooxypropanoic acid
SMIL N[C@@H](COP(=O)(O)O)C(=O)O

Tekniske data

Kemisk navn eller materiale L-O-Phosphoserine
InChI formel InChI=1S/C3H8NO6P/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)/t2-/m0/s1
Mængde 1 g
Formel vægt 185.0089
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.