Learn More
l-Norvaline, Sigma-Aldrich™
Product Code. 15802181
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
15802181 1 g 1g
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15802181 Supplier Sigma Aldrich Supplier No. 8415050001
 Restricted Item
Forventet forsendelse: 25-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
Melting Point >310°C
CAS 6600-40-4
MDL Number MFCD00064421
Synonym L-Norvaline, L-Norvaline
Molecular Weight (g/mol) 117.15
Quantity 1 g
Grade Research