Få mere at vide
L-Norleucine, TRC
Artikelnummer. 30006044
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006044 1 g
30010580 250 mg
30003457 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006044 Leverandør Toronto Research Chemicals Leverandørnr. TRCN7120001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 327-57-1
Molekylær formel C6 H13 N O2
Molekylvægt (g/mol) 131.17
Synonym L-Norleucine, Norleucine, L- (8CI), (S)-2-Aminohexanoic acid, (S)-Norleucine, (S)-α-Aminohexanoic acid, 2-Aminocaproic acid, 2-Aminohexanoic acid, Caprine, Glycoleucine, Hexanoic acid, 2-amino-, (S)-, L-(+)-Norleucine, L-2-Aminohexanoic acid, NSC 10378, NSC 74430, Norleucine, α-Aminocaproic acid
IUPAC navn (2S)-2-aminohexanoic acid
SMIL CCCC[C@H](N)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Norleucine (Aminocaproic acid)
Renhedsgrad noter HPLC
InChI formel InChI=1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m0/s1
Mængde 1 g
Formel vægt 131.0946
Procent renhed >95%