Få mere at vide
L-Norleucine-d9, TRC
Artikelnummer. 30419472
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30419472 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419472 Leverandør Toronto Research Chemicals Leverandørnr. TRCN7120021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Norleucine-3,3,4,4,5,5,6,6,6-d9
CAS 1331889-36-1
Molekylær formel C6 2H9 H4 N O2
Mængde 1 mg
Synonym (2S)-2-Amino-3,3,4,4,5,5,6,6,6-nonadeuteriohexanoic acid, L-Norleucine-d9, L-Norleucine-3,3,4,4,5,5,6,6,6-D9
InChI formel InChI=1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m0/s1/i1D3,2D2,3D2,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[C@H](N)C(=O)O
IUPAC navn (2S)-2-amino-3,3,4,4,5,5,6,6,6-nonadeuteriohexanoic acid
Molekylvægt (g/mol) 140.23
Formel vægt 140.1511