missing translation for 'onlineSavingsMsg'
Learn More

L-Methionyl diphenylborinate, 95%, Thermo Scientific™

Catalog Number 15422168
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
116628-54-7
C17H20BNO2S
313.222
MFCD22380723
LXKKLABPVUXKKX-INIZCTEOSA-N
l-methionyl diphenylborinate, diphenylboranyl 2s-2-amino-4-methylsulfanyl butanoate, diphenylborinic acid l-methionyl ester
73996184
diphenylboranyl (2S)-2-amino-4-methylsulfanylbutanoate
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CCSC)N
This item is not returnable. View return policy
116628-54-7
C17H20BNO2S
313.222
MFCD22380723
LXKKLABPVUXKKX-INIZCTEOSA-N
l-methionyl diphenylborinate, diphenylboranyl 2s-2-amino-4-methylsulfanyl butanoate, diphenylborinic acid l-methionyl ester
73996184
diphenylboranyl (2S)-2-amino-4-methylsulfanylbutanoate
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CCSC)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 313.22
Percent Purity 95%
Chemical Name or Material L-Methionyl diphenylborinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'