Få mere at vide
L-Methionine-S-methyl Sulfonium Chloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-Methionine-S-methyl Sulfonium Chloride |
| CAS | 1115-84-0 |
| Molekylær formel | C6 H14 N O2 S . Cl |
| Mængde | 500 mg |
| Synonym | Sulfonium, [(3S)-3-amino-3-carboxypropyl]dimethyl-, chloride (1:1), Sulfonium, (3-amino-3-carboxypropyl)dimethyl-, chloride, (S)-, Sulfonium, (3-amino-3-carboxypropyl)dimethyl-, chloride, L- (8CI), Sulfonium, [(3S)-3-amino-3-carboxypropyl]dimethyl-, chloride (9CI), Vitamin U (6CI,7CI), Ardesyl, Cabagin, Cabagin U, Epadyn U, L-Methionine methylsulfonium chloride, L-Methylmethionine sulfonium chloride, MMSC, Merastom, Methionine-S-methyl sulfonium chloride, Methylmethioninesulfonium chloride, S-Methylmethionine chloride, S-Methylmethionine sulfonium chloride, U-Vit, Vitas U, Yucron, L-Methionine-S-methyl sulfonium chloride |
| InChI formel | InChI=1S/C6H13NO2S.ClH/c1-10(2)4-3-5(7)6(8)9;/h5H,3-4,7H2,1-2H3;1H/t5-;/m0./s1 |
| SMIL | [Cl-].C[S+](C)CC[C@H](N)C(=O)O |
| IUPAC navn | [(3S)-3-amino-3-carboxypropyl]-dimethylsulfanium;chloride |
| Molekylvægt (g/mol) | 199.7 |
| Formel vægt | 199.0434 |