Få mere at vide
L-Methionine-S-methyl Sulfonium Chloride-d6 Hydrochloride, TRC
Artikelnummer. 30482335
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30482335 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482335 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26047225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Methionine-S-methyl Sulfonium Chloride-d6 Hydrochloride
CAS 362049-54-5
Molekylær formel C6 D6 H8 N O2 S . Cl
Mængde 25 mg
Synonym Sulfonium, [(3S)-3-amino-3-carboxypropyl]di(methyl-d3)-, chloride (9CI), L-Methionine-S-methyl Sulfonium Chloride-d6, L-Methionine-S-methyl Sulfonium Chloride-D6 (S,S-dimethyl D6)
InChI formel InChI=1S/C6H13NO2S.ClH/c1-10(2)4-3-5(7)6(8)9;/h5H,3-4,7H2,1-2H3;1H/t5-;/m0./s1/i1D3,2D3;
SMIL [Cl-].[2H]C([2H])([2H])[S+](CC[C@H](N)C(=O)O)C([2H])([2H])[2H]
IUPAC navn [(3S)-3-amino-3-carboxypropyl]-bis(trideuteriomethyl)sulfanium;chloride
Molekylvægt (g/mol) 205.736
Formel vægt 205.081