Få mere at vide
L-Methionine-3,3,4,4-d4, TRC
Artikelnummer. 30471703
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30471703 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471703 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26044110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Methionine-3,3,4,4-d4
CAS 67866-75-5
Molekylær formel C5 D4 H7 N O2 S
Mængde 10 mg
Synonym L-Methionine-3,3,4,4-d4 (9CI)
InChI formel InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i2D2,3D2
SMIL [2H]C([2H])(SC)C([2H])([2H])[C@H](N)C(=O)O
IUPAC navn (2S)-2-amino-3,3,4,4-tetradeuterio-4-methylsulfanylbutanoic acid
Molekylvægt (g/mol) 153.236
Formel vægt 153.0762