Få mere at vide
L-Lyxose, TRC
Artikelnummer. 30467964
Skift visning
Mængde:
1 g
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30467964 5 g -
30477414 1 g -
30484037 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30467964 Leverandør Toronto Research Chemicals Leverandørnr. TRCL5020005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1949-78-6
Molekylær formel C5 H10 O5
Molekylvægt (g/mol) 150.13
Synonym L-Lyxose, Lyxose, L- (8CI)
IUPAC navn (3R,4R,5S)-oxane-2,3,4,5-tetrol
SMIL O[C@H]1COC(O)[C@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale L-Lyxose
Mængde 5 g
InChI formel InChI=1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3+,4+,5?/m0/s1
Formel vægt 150.0528