Få mere at vide
L-Lysine-3,3,4,4,5,5,6,6-d8 Hydrochloride, TRC
Artikelnummer. 30480239
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30480239 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480239 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46891910MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Lysine-3,3,4,4,5,5,6,6-d8 Hydrochloride
CAS 344298-93-7
Molekylær formel C6 D8 H6 N2 O2 . Cl H
Mængde 10 mg
Synonym L-Lysine-3,3,4,4,5,5,6,6-d8, hydrochloride (1:1) (ACI), L-Lysine-3,3,4,4,5,5,6,6-d8, monohydrochloride (9CI), L-Lysine-3,3,4,4,5,5,6,6-D8 hydrochloride, L-Lysine-3,3,4,4,5,5,6,6-d8 hydrochloride, L-Lysine-d8 hydrochloride
InChI formel InChI=1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1/i1D2,2D2,3D2,4D2;
SMIL Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C([2H])([2H])[C@H](N)C(=O)O
IUPAC navn (2S)-2,6-diamino-3,3,4,4,5,5,6,6-octadeuteriohexanoic acid;hydrochloride
Molekylvægt (g/mol) 190.698
Formel vægt 190.132