Få mere at vide
L-Lysine-2,6,6-d3 HCl, TRC
Artikelnummer. 30480866
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30480866 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480866 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4689265MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Lysine-2,6,6-d3 HCl
CAS 2330878-43-6
Molekylær formel C6 D3 H11 N2 O2 . H Cl
Mængde 5 mg
Synonym L-Lysine-2,6,6-d3 HCl, L-Lysine-2,6,6-d3 hydrochloride, L-Lysine-2,6,6-d3 monohydrochloride, L-Lysine-2,6,6-D3 hydrochloride, (2S)-2,6-Diamino-2,6,6-trideuteriohexanoic acid monohydrochloride
InChI formel InChI=1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1/i4D2,5D;
SMIL Cl.[2H]C([2H])(N)CCC[C@]([2H])(N)C(=O)O
IUPAC navn (2S)-2,6-diamino-2,6,6-trideuteriohexanoic acid;hydrochloride
Molekylvægt (g/mol) 185.667
Formel vægt 185.101