Få mere at vide
L-Lysine-15N Dihydrochloride Salt, TRC
Artikelnummer. 30008268
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30008268 5 mg -
30003991 25 mg -
30007204 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008268 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4689255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 204451-50-3
Molekylær formel C6H14N15NO2 • 2(HCl)
Molekylvægt (g/mol) 147.1823646
Synonym L-Lysine Dihydrochloride α-15N
SMIL NCCCC[C@H]([15NH2])C(O)=O.Cl.Cl
Tekniske data
Kemisk navn eller materiale L-Lysine-15N Dihydrochloride Salt
Mængde 5 mg
InChI formel InChI=1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)/t5-/m0/s1/i8+1