Få mere at vide
L-Leucine-d3-N-FMOC (methyl-d3), CDN
Artikelnummer. 30312436
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312436 0.5 g 0.5g
30318596 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312436 Leverandør CDN Leverandørnr. CDND57330.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-5,5,5-D3
CAS 538372-74-6
Molekylær formel C21 D3 H20 N O4
Mængde 0.5 g
Synonym L-Leucine-5,5,5-d3, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (9 CI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-5,5,5-D3, L-Leucine-d3-N-FMOC (methyl-d3), (2 S)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-methylpentanoic-5,5,5-d3 acid, N-9-Fluorenylmethoxycarbonyl-L-leucine-d3 (methyl-d3), N-FMOC-L-leucine-5,5,5-d3, FMOC-L-Leucine-d3
InChI formel InChI=1 S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19 H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m0/s1/i1D3/t13?,19-
SMIL [2 H]C([2 H])([2 H])C(C)C[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC navn (2 S)-5,5,5-trideuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
Molekylvægt (g/mol) 356.4301
Formel vægt 356.1815 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity