missing translation for 'onlineSavingsMsg'
Få mere at vide

L-Leucine-3,3-d2, CDN

Artikelnummer. 30326164
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326164 0.05 g
30315371 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326164 Leverandør CDN Leverandørnr. CDND54820.05G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale L-Leucine-3,3-D2
CAS 362049-59-0
Molekylær formel C6 D2 H11 N O2
Mængde 0.05 g
Synonym L-Leucine-3,3-d2 (9 CI, ACI), L-Leucine-β,β-d2, L-Leucine-d2, (S)-2-Amino-(3,3-dideuterio)-4-methylpentanoic acid, (2 S)-4-Methyl-2-aminopentanoic-3,3-d2 acid, L-Leucine-3,3-D2
InChI formel InChI=1 S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i3D2
SMIL [2 H]C([2 H])(C(C)C)[C@H](N)C(=O)O
IUPAC navn (2 S)-2-amino-3,3-dideuterio-4-methylpentanoic acid
Molekylvægt (g/mol) 133.1852
Formel vægt 133.1072 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.