Få mere at vide
L-Leucine-3,3-d2, CDN
Artikelnummer. 30315371
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315371 0.1 g
30326164 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315371 Leverandør CDN Leverandørnr. CDND54820.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Leucine-3,3-D2
CAS 362049-59-0
Molekylær formel C6 D2 H11 N O2
Mængde 0.1 g
Synonym L-Leucine-3,3-d2 (9 CI, ACI), L-Leucine-β,β-d2, L-Leucine-d2, (S)-2-Amino-(3,3-dideuterio)-4-methylpentanoic acid, (2 S)-4-Methyl-2-aminopentanoic-3,3-d2 acid, L-Leucine-3,3-D2
InChI formel InChI=1 S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i3D2
SMIL [2 H]C([2 H])(C(C)C)[C@H](N)C(=O)O
IUPAC navn (2 S)-2-amino-3,3-dideuterio-4-methylpentanoic acid
Molekylvægt (g/mol) 133.1852
Formel vægt 133.1072 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity